3,3'-Diaminobenzidine (DAB), Sigma-Aldrich®
Supplier: Merck
Synonyms: [1,1'-Biphenyl]-3,3',4,4'-tetraamine, DAB, 3,3',4,4'-Biphenyltetramin, Biphenyl-3,3',4,4'-tetrayltetraamine
M-Clarity™ quality level = MQ200
D12384-1G D12384-25G D12384-5G
SIALD12384-1GEA 37.2 EUR
SIALD12384-1G SIALD12384-25G SIALD12384-5G
3,3'-Diaminobenzidine (DAB), Sigma-Aldrich®
3,3'-Diaminobenzidine
Formula: C₁₂H₁₄N₄
MW: 214.27 g/mol
MDL Number: MFCD00007725
CAS Number: 91-95-2
EINECS: 202-110-6