3',5'-Bis(trifluoromethyl)acetophenone

Supplier: Thermo Scientific

Synonyms: 1,3-Di(trifluormethyl)-5-ethanoylbenzol, 1-(3,5-Bis(trifluoromethyl)phenyl)ethanone, 1-[3,5-Bis(trifluoromethyl)phenyl]ethan-1-one

RF03488.1 RF03488.10 RF03488.50
MAYBRF03488.1EA 0 EUR
MAYBRF03488.1 MAYBRF03488.10 MAYBRF03488.50
3',5'-Bis(trifluoromethyl)acetophenone
3',5'-Bis(trifluoromethyl)acetophenone
Formula: C₁₀H₆F₆O
MW: 256.15 g/mol
Boiling Pt: 95 °C (15 mmHg)
Density: 1,42 g/cm³ (20 °C)
MDL Number: MFCD00009910
CAS Number: 30071-93-3
EINECS: 250-023-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR