1,3-Diaminopropane, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1,3-Propanediamine, Trimethylenediamine
D23602-25G
D23602-500G
D23602-3KG
SIALD23602-25GEA
35.3
EUR
SIALD23602-25G
SIALD23602-500G
SIALD23602-3KG
1,3-Diaminopropane, Sigma-Aldrich®
1,3-Diaminopropane
Formula:
C₃H₁₀N₂ MW: 74.13 g/mol Boiling Pt: 140 °C (1013 hPa) Melting Pt: –12 °C Density: 0.863 g/cm³ (20 °C) Flash Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008228 CAS Number: 109-76-2 EINECS: 203-702-7 UN: 2734 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...