1,3-Difluoro-4-nitrobenzene
Supplier: Thermo Scientific
Synonyms: 2,4-Difluoro-1-nitrobenzene, 2,4-Difluoronitrobenzene
SB00742.1 SB00742.10 SB00742.50
MAYBSB00742.1EA 0 EUR
MAYBSB00742.1 MAYBSB00742.10 MAYBSB00742.50
1,3-Difluoro-4-nitrobenzene
1,3-Difluoro-4-nitrobenzene
Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 203
Melting Pt: 10
Density: 1.4571 g/cm³ (14 °C)
MDL Number: MFCD00007050
CAS Number: 446-35-5
EINECS: 207-167-0
UN: 2810
ADR: 6.1 III