1,4-Diaminobutane dihydrochloride

Supplier: EHRENSTORFER GMBH

EHERC16584000EA 79.4 EUR
EHERC16584000
1,4-Diaminobutane dihydrochloride
1,4-Diaminobutane dihydrochloride
Formula: C₄H₁₄Cl₂N₂
MW: 161,07 g/mol
Density: 1,31 g/cm³ (20 °C)
MDL Number: MFCD00012526
CAS Number: 333-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR