2',5'-Dihydroxyacetophenone
Supplier: Thermo Scientific
Synonyms:
1-(2,5-Dihydroxyphenyl)ethan-1-one, 1-(2,5-Dihydroxyphenyl)ethanone, 2,5-Dihydroxyacetophenone
SB01933.1
SB01933.10
SB01933.50
MAYBSB01933.1EA
0
EUR
MAYBSB01933.1
MAYBSB01933.10
MAYBSB01933.50
2',5'-Dihydroxyacetophenone
2',5'-Dihydroxyacetophenone
Formula:
C₈H₈O₃ Boiling Pt: 296 °C (1013 hPa) Melting Pt: 199…202 °C |
MDL Number:
MFCD00002343 CAS Number: 490-78-8 EINECS: 207-716-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...