DL-Camphor

Supplier: Roth Carl

ROTH7552.1EA 123 EUR
ROTH7552.1
DL-Camphor
DL-Camphor
Formula: C₁₀H₁₆O
MW: 152,24 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: ∼180 °C
Density: 0,992 g/cm³ (25 °C)
Flash Pt: 66 °C
MDL Number: MFCD00074738
UN: 2717
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR