Prednisone, Sigma-Aldrich®

Supplier: Merck


MQ200

P6254-10G P6254-1G P6254-25G P6254-5G
SIALP6254-10GEA 237 EUR
SIALP6254-10G SIALP6254-1G SIALP6254-25G SIALP6254-5G
Prednisone, Sigma-Aldrich®
Prednisone
Formula: C₂₁H₂₆O₅
MW: 358.43 g/mol
Melting Pt: 230…232 °C
MDL Number: MFCD00003608
CAS Number: 53-03-2
EINECS: 200-160-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR