2-Amino-5-chlorobenzophenone 98%
Supplier: Thermo Fisher Scientific
Warning
103341000 103340250 103345000
ACRO103341000EA 39.4 EUR
ACRO103341000 ACRO103340250 ACRO103345000
2-Amino-5-chlorobenzophenone 98%
2-Amino-5-chlorobenzophenone
Formula: C₁₃H₁₀ClNO
Boiling Pt: 207 °C (760 mmHg)
Melting Pt: 96…100 °C
Flash Pt: 211 °C
Storage Temperature: Ambient
MDL Number: MFCD00007839
CAS Number: 719-59-5
EINECS: 211-949-7
Order Now
Specification Test Results
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 100 °C
GC ≥97.5 %
Titration with HClO4 ≥97.5 %