2-Amino-5-chlorobenzophenone ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-Amino-4-chloro-2-(phenylmethanoyl)benzene, (2-Amino-5-chlorophenyl)(phenyl)methanone
A15720.0E A15720.22 A15720.36
A15720.0EEA 534 EUR
A15720.0E A15720.22 A15720.36
2-Amino-5-chlorobenzophenone ≥98%
2-Amino-5-chlorobenzophenone
Purity: 98+% CAS No.: 719-59-5 Molecular Formula: C13H10ClNO Formula Weight: 231.68 Flash Point: 211°(412°F) Melting Point: 96-99° Beilstein Reference: 475640 MDL No.: MFCD00007839
Formula: C₁₃H₁₀ClNO
MW: 231.68 g/mol
Boiling Pt: 424
Melting Pt: 96…99 °C
Density: 1.33
Flash Pt: 211 °C (412 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007839
CAS Number: 719-59-5
EINECS: 211-949-7