2-Amino-5-chlorobenzophenone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Amino-4-chloro-2-(phenylmethanoyl)benzene, (2-Amino-5-chlorophenyl)(phenyl)methanone

M-Clarity™ quality level = MQ100

A45564-100G A45564-25G
SIALA45564-100GEA 60.5 EUR
SIALA45564-100G SIALA45564-25G
2-Amino-5-chlorobenzophenone, Sigma-Aldrich®
2-Amino-5-chlorobenzophenone
Formula: C₁₃H₁₀ClNO
MW: 231.68 g/mol
Boiling Pt: 424 °C (1013 hPa)
Melting Pt: 96…99 °C
MDL Number: MFCD00007839
CAS Number: 719-59-5
EINECS: 211-949-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR