4'-Aminoacetophenone
Supplier: Thermo Scientific
Warning
Synonyms: 1-(4-Aminophenyl)ethanone, p-Aminoacetophenone
SB01175.1 SB01175.10 SB01175.50
MAYBSB01175.1EA 0 EUR
MAYBSB01175.1 MAYBSB01175.10 MAYBSB01175.50
4'-Aminoacetophenone
4'-Aminoacetophenone
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 101…103 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007896
CAS Number: 99-92-3
EINECS: 202-801-2
Merck Index: 13,00412