1-Bromo-2-(2-methoxyethoxy)ethane, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Bromo-3,6-dioxaheptane, 2-(2-Methoxyethoxy)ethyl bromide, m-PEG2-Br

M-Clarity™ quality level = MQ100

639842-10G 639842-50G
SIAL639842-10GEA 243 EUR
SIAL639842-10G SIAL639842-50G
1-Bromo-2-(2-methoxyethoxy)ethane, Sigma-Aldrich®
1-Bromo-2-(2-methoxyethoxy)ethane
Formula: BrH₂CCH₂OCH₂CH₂OCH₃
MW: 183.05 g/mol
Boiling Pt: 172 °C (1013 hPa)
Melting Pt: –23 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000238
CAS Number: 54149-17-6
EINECS: 259-000-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR