1-Bromo-2-methylnaphthalene, Sigma-Aldrich®

Supplier: Merck
Warning

124109-100G 124109-25G
SIAL124109-100GEA 558 EUR
SIAL124109-100G SIAL124109-25G
1-Bromo-2-methylnaphthalene, Sigma-Aldrich®
1-Bromo-2-methylnaphthalene
Formula: C₁₁H₉Br
MW: 221.1 g/mol
Boiling Pt: 296 °C (1013 hPa)
Density: 1,41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003871
CAS Number: 2586-62-1
EINECS: 219-966-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR