4'-Bromoacetophenone
Supplier: Thermo Scientific
Warning
Synonyms: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
SB01774.1 SB01774.10 SB01774.50
MAYBSB01774.1EA 0 EUR
MAYBSB01774.1 MAYBSB01774.10 MAYBSB01774.50
4'-Bromoacetophenone
4'-Bromoacetophenone
Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 49…52 °C
Density: 1.736 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000105
CAS Number: 99-90-1
EINECS: 202-799-3
UN: 2811
ADR: 6.1,II
Merck Index: 13,01387