2'-Chloro-5'-(trifluoromethyl)acetophenone ≥97%
Supplier: Thermo Fisher Scientific
Warning
B23524.03 B23524.06
B23524.03EA 35.8 EUR
B23524.03 B23524.06
2'-Chloro-5'-(trifluoromethyl)acetophenone ≥97%
2'-Chloro-5'-(trifluoromethyl)acetophenone
Purity: 97+% CAS No.: 71648-45-8 Molecular Formula: C9H6ClF3O Formula Weight: 222.60 MDL No.: MFCD00045092
Formula: C₉H₆ClF₃O
MW: 222.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045092
CAS Number: 71648-45-8