2-Chlorophenol ≥99%, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Chloro-2-hydroxybenzene
185779-100G 185779-500G 185779-5G
SIAL185779-100GEA 54.2 EUR
SIAL185779-100G SIAL185779-500G SIAL185779-5G
2-Chlorophenol ≥99%, Sigma-Aldrich®
2-Chlorophenol
Formula: C₆H₅ClO
MW: 128.56 g/mol
Boiling Pt: 175
Melting Pt: 9 °C
Density: 1.25 g/cm³ (20 °C)
Flash Pt: 72
Storage Temperature: Ambient
MDL Number: MFCD00002159
CAS Number: 95-57-8
EINECS: 202-433-2
UN: 2021
ADR: 6.1 III
Merck Index: 12,02173