3-Chloro-4-methoxybenzonitrile, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

645575-1G 645575-5G
SIAL645575-1GEA 33.1 EUR
SIAL645575-1G SIAL645575-5G
3-Chloro-4-methoxybenzonitrile, Sigma-Aldrich®
3-Chloro-4-methoxybenzonitrile
Formula: C₈H₆ClNO
MW: 167.59 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 107…109 °C
MDL Number: MFCD03093073
CAS Number: 102151-33-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR