4-Chlorothiophenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Chloro-4-mercaptobenzene, 4-Chlorobenzene-1-thiol, 4-Chlorobenzenethiol, p-Chlorothiophenol
125237-100G 125237-500G 125237-5G
SIAL125237-100GEA 95.4 EUR
SIAL125237-100G SIAL125237-500G SIAL125237-5G
4-Chlorothiophenol, Sigma-Aldrich®
4-Chlorothiophenol
Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 49…53 °C
Density: 1.224 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004847
CAS Number: 106-54-7
EINECS: 203-408-9
UN: 1759
ADR: 8,III