4-Fluoro-2-methoxybenzonitrile ≥97%
Supplier: Thermo Fisher Scientific
Danger
H26627.03 H26627.09
H26627.03EA 62.3 EUR
H26627.03 H26627.09
4-Fluoro-2-methoxybenzonitrile ≥97%
4-Fluoro-2-methoxybenzonitrile
Purity: 97% CAS No.: 191014-55-8 Molecular Formula: C8H6FNO Formula Weight: 151.14 Melting Point: 75-77° UN No.: 3439 MDL No.: MFCD04116335
Formula: C₈H₆FNO
MW: 151.14 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD04116335
CAS Number: 191014-55-8
EINECS: None
UN: 3439
ADR: 6.1 III