1-Dodecene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Dodec-1-ene
D221600-500G
D221600-100G
SIALD221600-500GEA
118
EUR
SIALD221600-500G
SIALD221600-100G
1-Dodecene, Sigma-Aldrich®
1-Dodecene
Formula:
C₁₂H₂₄ MW: 168.32 g/mol Boiling Pt: 213 °C (1013 hPa) Melting Pt: –35 °C Density: 0,7582 g/cm³ (20 °C) Flash Pt: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008961 CAS Number: 112-41-4 EINECS: 203-968-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...