3'-Ethylacetophenone

Supplier: Thermo Scientific

TL00574DA TL00574EA
MAYBTL00574DAEA 0 EUR
MAYBTL00574DA MAYBTL00574EA
3'-Ethylacetophenone
3'-Ethylacetophenone
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00009660
CAS Number: 22699-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR