4'-Fluoroacetophenone
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
SB00895.1
SB00895.10
SB00895.50
MAYBSB00895.1EA
0
EUR
MAYBSB00895.1
MAYBSB00895.10
MAYBSB00895.50
4'-Fluoroacetophenone
4'-Fluoroacetophenone
Formula:
C₈H₇FO MW: 138.14 g/mol Boiling Pt: 196 °C (1013 hPa) Density: 1,138 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000354 CAS Number: 403-42-9 EINECS: 206-960-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...