5-Fluoro-2-nitrotoluene
Supplier: Thermo Scientific
Warning
Synonyms: 1-Fluoro-3-methyl-4-nitrobenzene
SB01285.1 SB01285.10 SB01285.50
MAYBSB01285.1EA 0 EUR
MAYBSB01285.1 MAYBSB01285.10 MAYBSB01285.50
5-Fluoro-2-nitrotoluene
5-Fluoro-2-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 218
Melting Pt: 28
Density: 1.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007283
CAS Number: 446-33-3
EINECS: 207-164-4
UN: 2810
ADR: 6.1 III