1-Isoquinolinecarboxylic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Isoquinoline-1-carboxylic acid
M-Clarity™ quality level = MQ100
150134-25G
150134-5G
SIAL150134-25GEA
215
EUR
SIAL150134-25G
SIAL150134-5G
1-Isoquinolinecarboxylic acid, Sigma-Aldrich®
1-Isoquinolinecarboxylic acid
Formula:
C₁₀H₇NO₂ MW: 173.17 g/mol Boiling Pt: 345 °C (1013 hPa) Melting Pt: 161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006901 CAS Number: 486-73-7 EINECS: 207-639-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...