2-Nitro-4-(trifluoromethyl)anisole
Supplier: Thermo Scientific
Synonyms:
4-Methoxy-3-nitrobenzotrifluoride, 1-Methoxy-2-nitro-4-(trifluoromethyl)benzene
BTB03145.1
BTB03145.10
BTB03145.50
MAYBBTB03145.1EA
0
EUR
MAYBBTB03145.1
MAYBBTB03145.10
MAYBBTB03145.50
2-Nitro-4-(trifluoromethyl)anisole
2-Nitro-4-(trifluoromethyl)anisole
Formula:
C₈H₆F₃NO₃ MW: 221.14 g/mol Boiling Pt: 90…95 °C (0.5 mmHg) Melting Pt: 47 °C |
MDL Number:
MFCD00007099 CAS Number: 394-25-2 EINECS: 206-891-4 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...