1-(2-Pyridylazo)-2-naphthol (PAN), Sigma-Aldrich®
Supplier: Merck
Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol
M-Clarity™ quality level = MQ200
101036-25G 101036-5G
SIAL101036-25GEA 396 EUR
SIAL101036-25G SIAL101036-5G
1-(2-Pyridylazo)-2-naphthol (PAN), Sigma-Aldrich®
1-(2-Pyridylazo)-2-naphthol
Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Boiling Pt: 412 °C (1013 hPa)
Melting Pt: 137…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00004071
CAS Number: 85-85-8
EINECS: 201-637-9