2-(Chloromethyl)benzimidazole 95%
Supplier: Thermo Scientific
Warning
Synonyms: 2-(Chloromethyl)-1H-benzimidazole
109390050 109390250
ACRO109390050EA 37.4 EUR
ACRO109390050 ACRO109390250
2-(Chloromethyl)benzimidazole 95%
2-(Chloromethyl)benzimidazole
Formula: C₈H₇ClN₂
MW: 166.61 g/mol
Melting Pt: 145…150 °C
MDL Number: MFCD00005602
CAS Number: 4857-04-9
EINECS: 225-452-8
Order Now
Specification Test Results
Appearance White to yellow to brown Powder
Infrared spectrum Conforms
Melting point 143 to 150 °C
HPLC ≥94 %
Titration with HClO4 94 to 106 %