2,2,2-Trifluorothanesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2,2,2-Trifluoroethanesulphonyl chloride, (2,2,2-Trifluoroethyl)sulphonyl chloride
324787-1G
324787-250MG
324787-5G
SIAL324787-1GEA
280
EUR
SIAL324787-1G
SIAL324787-250MG
SIAL324787-5G
2,2,2-Trifluorothanesulphonyl chloride, Sigma-Aldrich®
2,2,2-Trifluorothanesulphonyl chloride
Formula:
C₂H₂ClF₃O₂S MW: 182.55 g/mol Boiling Pt: 135 °C (1013 hPa) Density: 1.6905 g/cm³ (23 °C) |
MDL Number:
MFCD00007458 CAS Number: 1648-99-3 EINECS: 216-713-7 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...