(±)-2,2-Dimethyl-3-hexanol ≥97%
Supplier: Thermo Scientific
Warning
B20244.06 B20244.14
B20244.06EA 75.2 EUR
B20244.06 B20244.14
(±)-2,2-Dimethyl-3-hexanol ≥97%
(±)-2,2-Dimethyl-3-hexanol
Purity: 97% CAS No.: 4209-90-9 Molecular Formula: C8H18O Formula Weight: 130.23 MDL No.: MFCD00021929
Formula: C₈H₁₈O
MW: 130,23 g/mol
Density: 0.82
Storage Temperature: Ambient
MDL Number: MFCD00021929
CAS Number: 4209-90-9
EINECS: 224-135-1