2,5-Difluoro-4-hydroxybenzaldehyde 99
Supplier: Thermo Fisher Scientific
Warning
H31528.03 H31528.06
H31528.03EA 110 EUR
H31528.03 H31528.06
2,5-Difluoro-4-hydroxybenzaldehyde 99
2,5-Difluoro-4-hydroxybenzaldehyde
Purity: 99% CAS No.: 918523-99-6 Molecular Formula: C7H4F2O2 Formula Weight: 158.11 Melting Point: 145-148° MDL No.: MFCD04112521
Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Melting Pt: 145…148 °C
Storage Temperature: Ambient
MDL Number: MFCD04112521
CAS Number: 918523-99-6