2,6-Difluorobenzenesulphonamide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

542709-1G 542709-5G
SIAL542709-1GEA 45.4 EUR
SIAL542709-1G SIAL542709-5G
2,6-Difluorobenzenesulphonamide, Sigma-Aldrich®
2,6-Difluorobenzenesulphonamide
Formula: C₆H₅F₂NO₂S
MW: 193.17 g/mol
MDL Number: MFCD00729101
CAS Number: 60230-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR