2-Amino-3-hydroxybenzoic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Hydroxyanthranilic acid
M-Clarity™ quality level = MQ200
148776-1G 148776-250MG
SIAL148776-1GEA 1090 EUR
SIAL148776-1G SIAL148776-250MG
2-Amino-3-hydroxybenzoic acid, Sigma-Aldrich®
2-Amino-3-hydroxybenzoic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 357
Melting Pt: 164
MDL Number: MFCD00007700
CAS Number: 548-93-6
EINECS: 208-962-5