2-Aminocyclohexanol

Supplier: Thermo Scientific
Danger

Synonyms: 1-Amino-2-hydroxycyclohexane, 2-Aminocyclohexanol (cis- and trans- mixture)

BTB03899.1 BTB03899.10
MAYBBTB03899.1EA 0 EUR
MAYBBTB03899.1 MAYBBTB03899.10
2-Aminocyclohexanol
2-Aminocyclohexanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD00191368
CAS Number: 6850-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR