Methyl-2-chloroacetoacetate 95%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Chloroacetoacetic acid methyll ester, Methyl 2-chloroacetoacetate
179900100 179900500
ACRO179900100EA 89.9 EUR
ACRO179900100 ACRO179900500
Methyl-2-chloroacetoacetate 95%
Methyl 2-chloroacetoacetate
Formula: C₅H₇ClO₃
MW: 150.56 g/mol
Boiling Pt: 137 °C (1013 hPa)
Melting Pt: –33 °C
Density: 1.24 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00008757
CAS Number: 4755-81-1
EINECS: 225-286-6
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4455 to 1.4475 (20°C, 589 nm)
GC ≥94 %
HPLC ≥97.5 %