2-Chloro-6-methyl-5-nitropyridine 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 6-Chloro-2-methyl-3-nitropyridine, 2-Chloro-5-nitro-6-picoline, 6-Chloro-3-nitro-2-picoline
360820010 360820050
ACRO360820010EA 30.4 EUR
ACRO360820010 ACRO360820050
2-Chloro-6-methyl-5-nitropyridine 98%
2-Chloro-6-methyl-5-nitropyridine
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00234160
CAS Number: 22280-60-0
UN: 2811
ADR: 6.1 II
Order Now
Specification Test Results
Appearance Light beige to brown Powder
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥97.5 %