2-Chloroethanesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: (2-Chloroethyl)sulphonyl chloride
M-Clarity™ quality level = MQ200
302325-25G 302325-5G
SIAL302325-25GEA 106 EUR
SIAL302325-25G SIAL302325-5G
2-Chloroethanesulphonyl chloride, Sigma-Aldrich®
2-Chlorothanesulphonyl chloride
Formula: C₂H₄Cl₂O₂S
MW: 163.02 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.555 g/cm³ (20 °C)
MDL Number: MFCD00007461
CAS Number: 1622-32-8
EINECS: 216-594-1