2-Chloroimidazole ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Chloro-1H-imidazole

H51863.MD H51863.03
H51863.MDEA 103 EUR
H51863.MD H51863.03
2-Chloroimidazole ≥97%
2-Chloroimidazole

Purity: 97% CAS No.: 16265-04-6 Molecular Formula: C3H3ClN2 Formula Weight: 102.52 Melting Point: 165-166° MDL No.: MFCD02179530

Formula: C₃H₃ClN₂
MW: 102.52 g/mol
Melting Pt: 165…166 °C
Storage Temperature: Ambient
MDL Number: MFCD02179530
CAS Number: 16265-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR