1,1,1-Tris(hydroxymethyl)propane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
148083-3KG 148083-500G
SIAL148083-3KGEA 129 EUR
SIAL148083-3KG SIAL148083-500G
1,1,1-Tris(hydroxymethyl)propane, Sigma-Aldrich®
1,1,1-Tris(hydroxymethyl)propane
Formula: C₆H₁₄O₃
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004694
CAS Number: 77-99-6
EINECS: 201-074-9