2-Fluoro-5-methylbenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Fluoro-5-methylbenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

638889-1G 638889-5G
SIAL638889-1GEA 33.2 EUR
SIAL638889-1G SIAL638889-5G
2-Fluoro-5-methylbenzenesulphonyl chloride, Sigma-Aldrich®
2-Fluoro-5-methylbenzenesulphonyl chloride
Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
MDL Number: MFCD04039785
CAS Number: 870704-14-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR