2-Hydroxybenzimidazole, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Benzimidazolinone, 2(1H)-Benzimidazolone, 2-Hydroxy-1H-benzimidazole
M-Clarity™ quality level = MQ100
H19859-25G
H19859-5G
SIALH19859-25GEA
130
EUR
SIALH19859-25G
SIALH19859-5G
2-Hydroxybenzimidazole, Sigma-Aldrich®
2-Hydroxybenzimidazole
Formula:
C₇H₆N₂O MW: 134.14 g/mol Melting Pt: 320…322 °C Density: 1.42 g/cm³ (20 °C) |
MDL Number:
MFCD00127894 CAS Number: 615-16-7 EINECS: 210-412-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...