Toluene-2-sulphonamide
Supplier: Thermo Scientific
Warning
Synonyms: 2-Methylbenzene-1-sulphonamide, 2-Methylbenzenesulphonamide, o-Toluenesulphonamide
SB01480.1 SB01480.10 SB01480.50
MAYBSB01480.1EA 0 EUR
MAYBSB01480.1 MAYBSB01480.10 MAYBSB01480.50
Toluene-2-sulphonamide
Toluene-2-sulphonamide
Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 155…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007934
CAS Number: 88-19-7
EINECS: 201-808-8