2-Pentylfuran ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Amylfuran
A11607.09
A11607.18
A11607.30
A11607.09EA
54.3
EUR
A11607.09
A11607.18
A11607.30
2-Pentylfuran ≥98%
2-Pentylfuran
Purity: 98%
CAS No.: 3777-69-3
Molecular Formula: C9H14O
Formula Weight: 138.21
Storage Temperature: 2°C to 8°C
Flash Point: 45°(113°F)
Refractive Index: 1.4500
Boiling Point: 167°
Beilstein Reference: 107854
UN No.: 1993
MDL No.: MFCD00036497
Formula:
C₉H₁₄O Boiling Pt: 167 °C Density: 0.880 Flash Pt: 45 °C (113 °F) |
MDL Number:
MFCD00036497 CAS Number: 3777-69-3 EINECS: 223-234-7 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...