Baicalein ≥97%
Supplier: Thermo Scientific
Warning
Synonyms: 5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
L14185.MC L14185.ME
L14185.MCEA 45.3 EUR
L14185.MC L14185.ME
Baicalein ≥97%
Baicalein
Purity: 97% CAS No.: 491-67-8 Molecular Formula: C15H10O5 Formula Weight: 270.24 Melting Point: 268-270° Beilstein Reference: 272683 Merck Reference: 14,942 MDL No.: MFCD00017459
Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
Melting Pt: 268…270 °C
Storage Temperature: Ambient
MDL Number: MFCD00017459
CAS Number: 491-67-8
Merck Index: 14,942