3,4-Dibutoxy-3-cyclobutene-1,2-dione
Supplier: Thermo Scientific
Synonyms:
Squaric acid dibutylester, Dibutyl squarate, 3,4-Dibutoxycyclobut-3-ene-1,2-dione, 1,2-Dibutoxy-1-cyclobutene-3,4-dione
RDP00235.1
RDP00235.10
RDP00235EE
MAYBRDP00235.1EA
0
EUR
MAYBRDP00235.1
MAYBRDP00235.10
MAYBRDP00235EE
3,4-Dibutoxy-3-cyclobutene-1,2-dione
3,4-Dibutoxy-3-cyclobutene-1,2-dione
Formula:
C₁₂H₁₈O₄ MW: 226.27 g/mol Boiling Pt: 121 °C (1013 hPa) Density: 0.96 g/cm³ (20 °C) |
MDL Number:
MFCD00037150 CAS Number: 2892-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...