3,4-Dimethoxy-3-cyclobutene-1,2-dione, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,2-Dimethoxy-1-cyclobutene-3,4-dione, Dimethyl squarate, Squaric acid dimethylester
M-Clarity™ quality level = MQ100
377406-1G 377406-5G
SIAL377406-1GEA 27.2 EUR
SIAL377406-1G SIAL377406-5G
3,4-Dimethoxy-3-cyclobutene-1,2-dione, Sigma-Aldrich®
3,4-Dimethoxy-3-cyclobutene-1,2-dione
Formula: C₆H₆O₄
MW: 142.11 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 55 °C
MDL Number: MFCD00101316
CAS Number: 5222-73-1