3,5-Bis(trifluoromethyl)benzamide

Supplier: Thermo Scientific

BTB02664.1 BTB02664.10 BTB02664.50
MAYBBTB02664.1EA 0 EUR
MAYBBTB02664.1 MAYBBTB02664.10 MAYBBTB02664.50
3,5-Bis(trifluoromethyl)benzamide
3,5-Bis(trifluoromethyl)benzamide
Formula: C₉H₅F₆NO
MW: 257.14 g/mol
MDL Number: MFCD00015511
CAS Number: 22227-26-5
EINECS: 244-849-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR