3-Bromo-5-(trifluoromethyl)benzaldehyde ≥97%
Supplier: Thermo Scientific
Warning
Synonyms: 3-Bromo-5-trifluoromethylbenzaldehyde
H26197.03 H26197.06
H26197.03EA 32.1 EUR
H26197.03 H26197.06
3-Bromo-5-(trifluoromethyl)benzaldehyde ≥97%
3-Bromo-5-(trifluoromethyl)benzaldehyde
Purity: 97% CAS No.: 477535-41-4 Molecular Formula: C8H4BrF3O Formula Weight: 253.02 Refractive Index: 1.5110
Formula: C₈H₄BrF₃O
MW: 253.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059506
CAS Number: 477535-41-4
EINECS: None