5-Bromo-2-chloro-3-nitropyridine 98%
Supplier: Thermo Fisher Scientific
Warning
379570050 379570250
ACRO379570050EA 24.9 EUR
ACRO379570050 ACRO379570250
5-Bromo-2-chloro-3-nitropyridine 98%
5-Bromo-2-chloro-3-nitropyridine
Formula: C₅H₂BrClN₂O₂
MW: 237.44 g/mol
Melting Pt: 64…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00222270
CAS Number: 67443-38-3
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 64 to 69 °C
HPLC ≥97.5 %