3-Cyano-5-methylpyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-Cyano-5-picoline, 5-Methylpyridine-3-carbonitrile, 5-Methylnicotinonitrile
M-Clarity™ quality level = MQ100
661929-1G
661929-250MG
SIAL661929-1GEA
105
EUR
SIAL661929-1G
SIAL661929-250MG
3-Cyano-5-methylpyridine, Sigma-Aldrich®
3-Cyano-5-methylpyridine
Formula:
C₇H₆N₂ MW: 118.14 g/mol Boiling Pt: 220 °C (1013 hPa) Melting Pt: 86 °C |
MDL Number:
MFCD00673151 CAS Number: 42885-14-3 EINECS: 255-988-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...